Please login to your online account to display your discounted pricing

Ciliobrevin A, Tocris™

Hedgehog (Hh) pathway antagonist; inhibits ciliogenesis

$223.30 - $904.70

Chemical Identifiers

Chemical Name or Material Ciliobrevin A
Assay Percent Range >98%
Molecular Formula C17H9Cl2N3O2
Formula Weight 358.18
Synonym 2, 4-Dichloro-α-(3, 4-4-oxo-2(1H)-quinazolinylidene)-β-oxobenzenepropanenitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

452910

 
r & d systems
452910
10mg Each for $223.30

452950

 
r & d systems
452950
50mg Each for $904.70
Description & Specifications

Specifications

Formula Weight 358.18
Quantity 10mg
Chemical Name or Material Ciliobrevin A
Assay Percent Range >98%
Molecular Formula C17H9Cl2N3O2
CAS 302803-72-1
Synonym 2, 4-Dichloro-α-(3, 4-4-oxo-2(1H)-quinazolinylidene)-β-oxobenzenepropanenitrile
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C