Please login to your online account to display your discounted pricing

Cilastatin sodium, Tocris

Dipeptidase inhibitor

$139.08 - $576.76

Chemical Identifiers

Chemical Name or Material Cilastatin sodium
Assay Percent Range >99%
Molecular Formula C16H25N2NaO5S
Formula Weight 380.43
Synonym (2Z)-7-[[(2R)-2-Amino-2-carboxyethyl]thio]-2-[[[(1S)-2, 2-dimethylcyclopropyl]carbonyl]amino]-2-heptenoic acid sodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

270910

 
r & d systems
RDS270910
10mg Each for $139.08

270950

 
r & d systems
RDS270950
50mg Each for $576.76
Description & Specifications

Specifications

Formula Weight 380.43
Quantity 10mg
Chemical Name or Material Cilastatin sodium
Assay Percent Range >99%
Molecular Formula C16H25N2NaO5S
CAS 81129-83-1
Synonym (2Z)-7-[[(2R)-2-Amino-2-carboxyethyl]thio]-2-[[[(1S)-2, 2-dimethylcyclopropyl]carbonyl]amino]-2-heptenoic acid sodium salt
Recommended Storage Desiccate at -20°C
Safety and Handling

Desiccate at -20°C