Please login to your online account to display your discounted pricing

Ciglitazone, Tocris

Selective PPARγ agonist

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material Ciglitazone
Assay Percent Range >99%
Molecular Formula C18H23NO3S
Formula Weight 333.44
Synonym 5-[[4-[(1-Methylcyclohexyl)methoxy]phenyl]methyl]-2, 4-thiazolidinedione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

130750

 
r & d systems
RDS130750
50mg Each for $976.15

130710

 
r & d systems
RDS130710
10mg Each for $239.63
Description & Specifications

Specifications

Chemical Name or Material Ciglitazone
CAS 74772-77-3
Recommended Storage Store at Room Temperature
Assay Percent Range >99%
Molecular Formula C18H23NO3S
Formula Weight 333.44
Synonym 5-[[4-[(1-Methylcyclohexyl)methoxy]phenyl]methyl]-2, 4-thiazolidinedione
Quantity 50mg
Safety and Handling

Store at Room Temperature