Ciglitazone, Tocris
Selective PPARγ agonist
Manufacturer:  R & D Systems RDS130750
 View more versions of this product
Catalog No. 130750
Specifications Description & Specifications
Specifications
Ciglitazone
>99%
333.44
50mg
74772-77-3
C18H23NO3S
5-[[4-[(1-Methylcyclohexyl)methoxy]phenyl]methyl]-2, 4-thiazolidinedione
Chemical Identifiers
>99%
333.44
Ciglitazone
C18H23NO3S
5-[[4-[(1-Methylcyclohexyl)methoxy]phenyl]methyl]-2, 4-thiazolidinedione
Safety and Handling
Recommended Storage : Store at Room Temperature