Please login to your online account to display your discounted pricing

Tocris™ CID 16020046

Selective GPR55 antagonist

$257.85 - $1,073.10

Specifications

Product Type CID 16020046
Inhibitors CID 16020046
Molecular Weight 425.44
Purity >99
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

495910R

 
tocris™
RDS495910
10mg Each for $257.85

495950R

 
tocris™
RDS495950
50mg Each for $1,073.10
Description & Specifications

Specifications

Product Type CID 16020046
Inhibitors CID 16020046
Molecular Weight 425.44
Purity >99

Selective GPR55 antagonist. Inhibits LPI-induced Ca2+ signaling (IC50 = 0.21μM in HEK-GPR55 cells), ERK1/2 phosphorylation and GPR55-mediated transcription factor activation. Displays weak inhibition of acetylcholinesterase,μ-opioid receptor, KCNH2 and hERG. Decreases LPI-induced GPR55 internalization. Reduces experimental intestinal inflammation in mice.

  • Chemical Name: 4-[4,6-Dihydro-4-(3-hydroxyphenyl)-3-(4-methylphenyl)-6-oxopyrrolo[3,4-c]pyrazol-5(1H)-yl]benzoic acid
  • Formula: C25H19N3O4
  • CAS Number: 834903-43-4
  • Solubility: Soluble to 100mM in DMSO
  • Purity: >99