Please login to your online account to display your discounted pricing

CI 994, Tocris

Histone deacetylase inhibitor

$175.83 - $734.99

Chemical Identifiers

Chemical Name or Material CI 994
Assay Percent Range >99%
Molecular Formula C15H15N3O2
Formula Weight 269.3
Synonym 4-(Acetylamino)-N-(2-aminophenyl)benzamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

295250

 
r & d systems
RDS295250
50mg Each for $734.99

295210

 
r & d systems
RDS295210
10mg Each for $175.83
Description & Specifications

Specifications

Formula Weight 269.3
Quantity 50mg
Chemical Name or Material CI 994
Assay Percent Range >99%
Molecular Formula C15H15N3O2
CAS 112522-64-2
Synonym 4-(Acetylamino)-N-(2-aminophenyl)benzamide
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C