missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ CI 994

$166.55 - $265.24

Chemical Identifiers

CAS 112522-64-2
Molecular Formula C15H15N3O2
Molecular Weight (g/mol) 269.304
MDL Number MFCD00866266
InChI Key VAZAPHZUAVEOMC-UHFFFAOYSA-N
Synonym tacedinaline, acetyldinaline, 4-acetamido-n-2-aminophenyl benzamide, 4-acetylamino-n-2'-aminophenyl benzamide, tacedinalina, ci994 tacedinaline, n-acetyldinaline, tacedinaline usan:inn, tacedinalina inn-spanish, 4-acetylamino-n-2-aminophenyl benzamide
PubChem CID 2746
IUPAC Name 4-acetamido-N-(2-aminophenyl)benzamide
SMILES CC(=O)NC1=CC=C(C=C1)C(=O)NC2=CC=CC=C2N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ65561MB View Documents Alfa Aesar™
J65561MB
25mg Each for $166.55
 
AAJ65561MC View Documents Alfa Aesar™
J65561MC
100mg Each for $265.24
 
Specifications

Chemical Identifiers

112522-64-2
269.304
VAZAPHZUAVEOMC-UHFFFAOYSA-N
2746
CC(=O)NC1=CC=C(C=C1)C(=O)NC2=CC=CC=C2N
C15H15N3O2
MFCD00866266
tacedinaline, acetyldinaline, 4-acetamido-n-2-aminophenyl benzamide, 4-acetylamino-n-2'-aminophenyl benzamide, tacedinalina, ci994 tacedinaline, n-acetyldinaline, tacedinaline usan:inn, tacedinalina inn-spanish, 4-acetylamino-n-2-aminophenyl benzamide
4-acetamido-N-(2-aminophenyl)benzamide

Specifications

Air Sensitive
25mg
C15H15N3O2
tacedinaline, acetyldinaline, 4-acetamido-n-2-aminophenyl benzamide, 4-acetylamino-n-2'-aminophenyl benzamide, tacedinalina, ci994 tacedinaline, n-acetyldinaline, tacedinaline usan:inn, tacedinalina inn-spanish, 4-acetylamino-n-2-aminophenyl benzamide
VAZAPHZUAVEOMC-UHFFFAOYSA-N
4-acetamido-N-(2-aminophenyl)benzamide
2746
Powder
White
112522-64-2
MFCD00866266
Soluble in DMSO
CC(=O)NC1=CC=C(C=C1)C(=O)NC2=CC=CC=C2N
269.304
269.3
CI 994
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

RTECSNumber : CU8702023

Recommended Storage : 2° to 8°C