Please login to your online account to display your discounted pricing

CI 966 hydrochloride, Tocris

Selective inhibitor of GAT-1

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material CI 966 hydrochloride
Assay Percent Range >98%
Molecular Formula C23H21F6NO3·HCl
Formula Weight 509.87
Synonym 1-[2-[bis[4-(Trifluoromethyl)phenyl]methoxy]ethyl]-1,2,5,6-tetrahydropyridine-3-carboxylic acid hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

129610

 
r & d systems
RDS129610
10mg Each for $239.63

129650

 
r & d systems
RDS129650
50mg Each for $976.15
Description & Specifications

Specifications

Formula Weight 509.87
Quantity 10mg
Chemical Name or Material CI 966 hydrochloride
Assay Percent Range >98%
Molecular Formula C23H21F6NO3·HCl
CAS 110283-66-4
Synonym 1-[2-[bis[4-(Trifluoromethyl)phenyl]methoxy]ethyl]-1,2,5,6-tetrahydropyridine-3-carboxylic acid hydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature