Please login to your online account to display your discounted pricing

CI 1020, Tocris

Highly selective, orally active ETA antagonist

$113.57 - $976.15

Chemical Identifiers

Chemical Name or Material CI 1020
Assay Percent Range >98%
Molecular Formula C28H26O9
Formula Weight 506.5
Synonym 3-(1,3-Benzodioxol-5-yl)-5-hydroxy-5-(4-methoxyphenyl)-4-[(3,4,5-trimethoxyphenyl)methyl]-2(5H)-furanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

29421

 
r & d systems
RDS29421
1mg Each for $113.57

294210

 
r & d systems
RDS294210
10mg Each for $239.63

294250

 
r & d systems
RDS294250
50mg Each for $976.15
Description & Specifications

Specifications

Formula Weight 506.5
Quantity 1mg
Assay Percent Range >98%
Molecular Formula C28H26O9
CAS 162256-50-0
Recommended Storage Store at -20°C
Synonym 3-(1,3-Benzodioxol-5-yl)-5-hydroxy-5-(4-methoxyphenyl)-4-[(3,4,5-trimethoxyphenyl)methyl]-2(5H)-furanone
Chemical Name or Material CI 1020
Safety and Handling

Store at -20°C