α-Chymotrypsin ≥40u/mg protein, MP Biochemicals™
Manufacturer:  Mp Biomedicals 0210046190
 View more versions of this product
Catalog No. ICN10046190
Description
Description
Specifications
Specifications
α-Chymotrypsin
500mg
9004-07-3
Powder
White
3X Crystallized from Bovine Pancreas, ≥40μ/mg protein
MFCD00130481
Chemical Identifiers
9004-07-3
MFCD00130481
Safety and Handling
Safety and Handling
Recommended Storage : Store at +4°C