missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

α-Chymotrypsin ≥40u/mg protein, MP Biochemicals™

Manufacturer:  Mp Biomedicals 0210046180

 View more versions of this product

Catalog No. ICN10046180



Specifications

Specifications

α-Chymotrypsin
0.1g
9004-07-3
Powder
White
3X Crystallized from Bovine Pancreas, ≥40μ/mg protein
MFCD00130481

Chemical Identifiers

9004-07-3
MFCD00130481
Safety and Handling

Safety and Handling

Recommended Storage : Store at +4°C

Documents