missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ CHUK (IKK α) Recombinant Human Protein
CHUK (IKK alpha) belongs to the cytokine activated protein complex that is an inhibitor of the NF-kappa B complex.
Supplier: Invitrogen™ PV4327
Description
CHUK (IKK alpha) is a serine/threonine kinase involved in signal transduction and cellular communication.
Specifications
NP_001269 | |
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton™ X-100, 2mM DTT, and50% Glycerol | |
1mg | |
RUO - research use only | |
CHUK (IKK α) | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
88.7kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
IKK Kinase Family | |
Recombinant | |
Baculovirus |