Please login to your online account to display your discounted pricing

Chrysophanic acid, 98%, ACROS Organics™


Quantity 50mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC211950500

 View more versions of this product

Catalog No. AC211950500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Solid
Formula Weight 254.24
Quantity 50mg
Color Yellow
Beilstein 08, 472
Merck Index 15, 2262
Melting Point 194° to 198°C
Chemical Name or Material Chrysophanic acid
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C15H10O4
MDL Number MFCD00001208
CAS 481-74-3
Synonym 1, 8-Dihydroxy-3-methylanthraquinone
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay Percent Range 98%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Chrysophanic acid
CAS 481-74-3
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C15H10O4
Formula Weight 254.24
MDL Number MFCD00001208
Synonym 1, 8-Dihydroxy-3-methylanthraquinone
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents