Please login to your online account to display your discounted pricing

Chrysin, 99+%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC110321000

 View more versions of this product

Catalog No. AC110321000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 254.24
Assay ≥99.0%
Physical Form Crystalline Powder
Quantity 100g
Color Yellow
Melting Point 284° to 290°C
Synonym 5, 7-Dihydroxyflavone
Chemical Name or Material Chrysin
Molecular Formula C15H10O4
CAS 480-40-0
Assay Percent Range 99+%
MDL Number MFCD00006834
Loss on Drying 1.0% max.
Packaging Glass bottle
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Chrysin
CAS 480-40-0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C15H10O4
Formula Weight 254.24
MDL Number MFCD00006834
Synonym 5, 7-Dihydroxyflavone