Please login to your online account to display your discounted pricing

Chrysanthemyl alcohol, 98%, mixture of cis and trans, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  186830010

 View more versions of this product

Catalog No. AC186830010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥97.5%
Formula Weight 154.25
Quantity 1g
Color Clear yellow
Chemical Name or Material Chrysanthemyl alcohol
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 98%
Refractive Index 1.4730 to 1.4750 (20°C, 589nm)
Assay Percent Range 98%
Molecular Formula C10H18O
MDL Number MFCD00001305
CAS 5617-92-5
Synonym 2, 2-Dimethyl-3-(2-methyl-1-propenyl)cyclopropanemethanol

Specifications

Chemical Name or Material Chrysanthemyl alcohol
Name Note 98%
CAS 5617-92-5
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H18O
Formula Weight 154.25
MDL Number MFCD00001305
Synonym 2, 2-Dimethyl-3-(2-methyl-1-propenyl)cyclopropanemethanol