Please login to your online account to display your discounted pricing

Chrysanthemyl alcohol, 98%, mixture of cis and trans, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Chrysanthemyl alcohol
Name Note 98%
CAS 5617-92-5
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C10H18O
Formula Weight 154.25g/mol
MDL Number MFCD00001305
Synonym 2, 2-Dimethyl-3-(2-methyl-1-propenyl)cyclopropanemethanol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC186830010

  SDS
thermo fisher scientific
186830010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4730 to 1.4750 (20°C, 589nm)
Color Clear yellow
Quantity 1g
Chemical Name or Material Chrysanthemyl alcohol
Name Note 98%
CAS 5617-92-5
Physical Form Clear yellow liquid
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C10H18O
Formula Weight 154.25g/mol
MDL Number MFCD00001305
Synonym 2, 2-Dimethyl-3-(2-methyl-1-propenyl)cyclopropanemethanol