Please login to your online account to display your discounted pricing

Chromotropic Acid Sodium Salt (Tech.), 60%, ACROS Organics™


Quantity 250g
Packaging Glass bottle

Manufacturer: fisher scientific acros  405262500

 View more versions of this product

Catalog No. AC405262500

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥55%
Purity Grade Technical Grade
Physical Form Powder
Formula Weight 342.27
Quantity 250g
Color Light gray to brown
EINECS Number 223-430-2
TSCA TSCA
Packaging Glass bottle
Chemical Name or Material Chromotropic acid sodium salt
Name Note tech., 60%
Assay Percent Range 60%
Molecular Formula C10H7NaO8S2
MDL Number MFCD00067682
CAS 3888-44-6
Synonym 4, 5-Dihydroxy-2, 7-naphthalenedisulfonic acid sodium salt
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Chromotropic acid sodium salt
Name Note tech., 60%
CAS 3888-44-6
Purity Grade Technical Grade
Assay ≥55%
Assay Percent Range 60%
Molecular Formula C10H7NaO8S2
Formula Weight 342.27
MDL Number MFCD00067682
Synonym 4, 5-Dihydroxy-2, 7-naphthalenedisulfonic acid sodium salt
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents