missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chromotropic Acid Sodium Salt (Tech.), 60%, ACROS Organics™

$245.79 - $245.79

Chemical Identifiers

CAS 3888-44-6
Molecular Formula C10H7NaO8S2
Molecular Weight (g/mol) 342.268
MDL Number MFCD00067682
InChI Key UPYOFRGSTDARPV-UHFFFAOYSA-M
Synonym 4, 5-Dihydroxy-2, 7-naphthalenedisulfonic acid sodium salt
PubChem CID 101979282
IUPAC Name sodium;8-hydroxy-3,6-disulfonaphthalen-1-olate
SMILES C1=C2C=C(C=C(C2=C(C=C1S(=O)(=O)O)[O-])O)S(=O)(=O)O.[Na+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC405262500 SDS View Documents Acros Organics
405262500
250g Each for $245.79
Specifications

Chemical Identifiers

3888-44-6
342.268
UPYOFRGSTDARPV-UHFFFAOYSA-M
101979282
C1=C2C=C(C=C(C2=C(C=C1S(=O)(=O)O)[O-])O)S(=O)(=O)O.[Na+]
C10H7NaO8S2
MFCD00067682
4, 5-Dihydroxy-2, 7-naphthalenedisulfonic acid sodium salt
sodium;8-hydroxy-3,6-disulfonaphthalen-1-olate

Specifications

3888-44-6
C10H7NaO8S2
4, 5-Dihydroxy-2, 7-naphthalenedisulfonic acid sodium salt
UPYOFRGSTDARPV-UHFFFAOYSA-M
sodium;8-hydroxy-3,6-disulfonaphthalen-1-olate
101979282
Powder
Technical
Brown to Gray
Chromotropic acid sodium salt, tech., 60%
60%
MFCD00067682
Authentic
C1=C2C=C(C=C(C2=C(C=C1S(=O)(=O)O)[O-])O)S(=O)(=O)O.[Na+]
342.268
342.27
60%
Glass bottle
250g
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 223-430-2

TSCA : TSCA