missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Chromotrope 2B
CAS: 548-80-1 | C16H9N3Na2O10S2 | 513.36 g/mol
$91.63 - $233.01
Chemical Identifiers
| CAS | 548-80-1 |
|---|---|
| Molecular Formula | C16H9N3Na2O10S2 |
| Molecular Weight (g/mol) | 513.36 |
| MDL Number | MFCD00003951 |
| InChI Key | MQHFURHHKNKIRN-GVLVODDDSA-L |
| Synonym | Acid Red 176; C.I. 16575 |
| PubChem CID | 44135472 |
| SMILES | [Na+].[Na+].OC1=CC(=CC2=C1C(=O)\C(=N/NC1=CC=C(C=C1)[N+]([O-])=O)C(=C2)S([O-])(=O)=O)S([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1133014
|
Thermo Scientific Chemicals
L1133014 |
25 g |
Each for $91.63
|
|
|||||
|
AAL1133022
|
Thermo Scientific Chemicals
L1133022 |
100 g |
Each for $233.01
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 548-80-1 | |
| 513.36 | |
| MQHFURHHKNKIRN-GVLVODDDSA-L | |
| 44135472 |
| C16H9N3Na2O10S2 | |
| MFCD00003951 | |
| Acid Red 176; C.I. 16575 | |
| [Na+].[Na+].OC1=CC(=CC2=C1C(=O)\C(=N/NC1=CC=C(C=C1)[N+]([O-])=O)C(=C2)S([O-])(=O)=O)S([O-])(=O)=O |
Specifications
| >300°C | |
| C16H9N3Na2O10S2 | |
| 4109733 | |
| Acid Red 176; C.I. 16575 | |
| [Na+].[Na+].OC1=CC(=CC2=C1C(=O)\C(=N/NC1=CC=C(C=C1)[N+]([O-])=O)C(=C2)S([O-])(=O)=O)S([O-])(=O)=O | |
| 513.36 | |
| 513.37 | |
| 25 g |
| 548-80-1 | |
| MFCD00003951 | |
| 14,2240 | |
| MQHFURHHKNKIRN-GVLVODDDSA-L | |
| disodium;5-hydroxy-3-[(4-nitrophenyl)hydrazinylidene]-4-oxonaphthalene-2,7-disulfonate | |
| 44135472 | |
| 97% | |
| Chromotrope 2B |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 208-959-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only