Please login to your online account to display your discounted pricing

Chromosulfuric acid, ACROS Organics™


Quantity 2.5L
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC295510025

 View more versions of this product

Catalog No. AC295510025

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Quantity 2.5L
Color Red-brown to dark brown
Density 1.8400g/mL
Packaging Glass bottle
Sulfuric Acid > 92%
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chromium (VI) Oxide > 1.3%
Chemical Name or Material Chromosulfuric acid
Molecular Formula Cr2K2O7·H2O4S
MDL Number MFCD00010952
CAS 65272-71-1
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
May cause an allergic skin reaction.
Causes serious eye damage.
May cause cancer.
May cause cancer by inhalation.

Specifications

Chemical Name or Material Chromosulfuric acid
CAS 65272-71-1
Molecular Formula Cr2K2O7·H2O4S
MDL Number MFCD00010952
Accessories

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
May cause an allergic skin reaction.
Causes serious eye damage.
May cause cancer.
May cause cancer by inhalation.

SDS & Safety Documents