Please login to your online account to display your discounted pricing

Alfa Aesar™ Chromostatin, Bovine

For cellular and molecular biology applications

Specifications

Product Type Chromostatin, bovine
Cross Reactivity Bovine
Molecular Weight 1954.00
Storage Requirements -30° to -10°C
Sequence Ser-Asp-Glu-Asp-Ser-Asp-Gly-Asp-Arg-Pro-Gln-Ala-Ser-Pro-Gly-Leu-Gly-Pro-Gly-Pro
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66182EXD

 
alfa aesar™
J66182EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66182MCR

 
alfa aesar™
J66182MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Chromostatin, bovine
Cross Reactivity Bovine
Molecular Weight 1954.00
Storage Requirements -30° to -10°C
Sequence Ser-Asp-Glu-Asp-Ser-Asp-Gly-Asp-Arg-Pro-Gln-Ala-Ser-Pro-Gly-Leu-Gly-Pro-Gly-Pro
Format Solid
Common Name Chromostatin-20
Regulatory Status RUO
Purification Method HPLC