Please login to your online account to display your discounted pricing

Chromone-3-carboxaldehyde, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  35574-0010

 View more versions of this product

Catalog No. AC355740010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 174.16
Quantity 1g
Color Orange
Melting Point 151° to 155°C
EINECS Number 241-451-5
Chemical Name or Material Chromone-3-carboxaldehyde
Infrared Spectrum Authentic
MDL Number MFCD00014667
CAS 17422-74-1
Health Hazard 1 AIR SENSITIVE
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 98%
Molecular Formula C10 H6 O3
Packaging Glass bottle

Specifications

Chemical Name or Material Chromone-3-carboxaldehyde
CAS 17422-74-1
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10 H6 O3
Formula Weight 174.16
MDL Number MFCD00014667
Accessories

AIR SENSITIVE

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.

SDS & Safety Documents