Learn More
Chromone-3-carbonitrile, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals A1568706
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Chromone-3-carbonitrile | |
173°C to 177°C | |
MFCD00052604 | |
UN3439 | |
SFWNPLLGXKJESA-UHFFFAOYSA-N | |
4-oxochromene-3-carbonitrile | |
521298 | |
97% |
50743-17-4 | |
C10H5NO2 | |
5 g | |
3-cyanochromone, chromone-3-carbonitrile, 4-oxo-4h-chromene-3-carbonitrile, 4-oxo-4h-1-benzopyran-3-carbonitrile, acmc-20an5h, maybridge1_006148, 4-oxo-chromene-3-carbonitrile, 3-cyano-4h-1-benzopyran-4-one, 4-oxo-1-benzopyran-3-carbonitrile | |
O=C1C(=COC2=CC=CC=C12)C#N | |
171.16 | |
171.16 |
Chemical Identifiers
50743-17-4 | |
171.16 | |
SFWNPLLGXKJESA-UHFFFAOYSA-N | |
521298 |
C10H5NO2 | |
MFCD00052604 | |
3-cyanochromone, chromone-3-carbonitrile, 4-oxo-4h-chromene-3-carbonitrile, 4-oxo-4h-1-benzopyran-3-carbonitrile, acmc-20an5h, maybridge1_006148, 4-oxo-chromene-3-carbonitrile, 3-cyano-4h-1-benzopyran-4-one, 4-oxo-1-benzopyran-3-carbonitrile | |
O=C1C(=COC2=CC=CC=C12)C#N |
Safety and Handling
GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures