missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Chromone-3-carbonitrile, 97%

Manufacturer:  Alfa Aesar™ A1568703

 View more versions of this product

Catalog No. AAA1568703



Specifications

Specifications

Chromone-3-carbonitrile
1g
C10H5NO2
UN3439
SFWNPLLGXKJESA-UHFFFAOYSA-N
4-oxochromene-3-carbonitrile
521298
97%
173°C to 177°C
50743-17-4
MFCD00052604
3-cyanochromone, chromone-3-carbonitrile, 4-oxo-4h-chromene-3-carbonitrile, 4-oxo-4h-1-benzopyran-3-carbonitrile, acmc-20an5h, 3-cyanochromone, maybridge1_006148, 4-oxo-chromene-3-carbonitrile, 3-cyano-4h-1-benzopyran-4-one, 4-oxo-1-benzopyran-3-carbonitrile
C1=CC=C2C(=C1)C(=O)C(=CO2)C#N
171.155
171.16

Chemical Identifiers

50743-17-4
171.155
SFWNPLLGXKJESA-UHFFFAOYSA-N
521298
C1=CC=C2C(=C1)C(=O)C(=CO2)C#N
C10H5NO2
MFCD00052604
3-cyanochromone, chromone-3-carbonitrile, 4-oxo-4h-chromene-3-carbonitrile, 4-oxo-4h-1-benzopyran-3-carbonitrile, acmc-20an5h, 3-cyanochromone, maybridge1_006148, 4-oxo-chromene-3-carbonitrile, 3-cyano-4h-1-benzopyran-4-one, 4-oxo-1-benzopyran-3-carbonitrile
4-oxochromene-3-carbonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

Documents