Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Chromogranin A, Rabbit Monoclonal Antibody

Obtain accurate and reproducible results in immunohistochemistry applications with Thermo Scientific™ Chromogranin A, Rabbit Monoclonal Antibody.

$303.33 - $921.31

Specifications

Antigen Chromogranin A
Clone SP12
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

RM9112R7

 
thermo scientific™ lab vision™
RM9112R7
7mL Each for $474.88

RM9112S

 
thermo scientific™ lab vision™
RM9112S
1mL Each for $921.31

RM9112S0

 
thermo scientific™ lab vision™
RM9112S0
100μL Each for $303.33

RM9112S1

 
thermo scientific
RM9112S1
500μL Each for $681.08
Description & Specifications

Specifications

Antigen Chromogranin A
Clone SP12
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Host Species Rabbit
Regulatory Status IVD
Research Discipline Oncogenesis
Type Primary
Immunogen Recombinant protein encoding human chromogranin A

Obtain accurate and reproducible results in immunohistochemistry applications with Thermo Scientific™ Chromogranin A, Rabbit Monoclonal Antibody, part of a comprehensive antibody portfolio. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.

Chromogranin A (a protein of 439 amino acid which is encoded on chromosome 14) is present in neuroendocrine cells throughout the body, including the neuroendocrine cells of the large and small intestine, adrenal medulla and pancreatic islets. It is an excellent marker for carcinoid tumors, phenochromocytomas, paragangliomas, and other neuroendocrine tumors. Coexpression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms.

Host Species: Rabbit

Clone: SP12

Isotype: IgG

Species Reactivity: Human. Others not known

Epitope: Not determined

Immunogen: Recombinant protein encoding human chromogranin A

Positive Control: Pancreas

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)