Learn More
Chromogranin A Ab-3 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Chromogranin A Ab-3, Mouse Monoclonal Antibody.
Supplier: Thermo Fisher Scientific MS382P1ABX
Description
Chromogranin A (a protein of 439-amino acid which is encoded on chromosome 14) is present in neuroendocrine cells throughout the body, including the neuroendocrine cells of the large and small intestine, adrenal medulla and pancreatic islets. It is an excellent marker for carcinoid tumors, pheo-chromocytomas, paragangliomas, and other neuro-endocrine tumors. Coexpression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms. Ab-3 cocktail is especially designed for sensitive detection of Chromogranin A in formalin-fixed, paraffin-embedded tissues.Host Species: Mouse
Clone: LK2H10 + PHE5
Isotype: IgG1 + IgG1
Species Reactivity: Human and Monkey. Others not known.
Immunogen: Human pheochromocytoma
Molecular Weight: 68-75 kDa
Positive Control: Pancreas
Cellular Localization: Cytoplasmic in endocrine cells
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
Chromogranin A Ab-3 | |
Monoclonal | |
1mg/mL | |
Mouse | |
100 μL | |
Oncogenesis | |
Human, Non-human Primate | |
IgG1 |
Immunohistochemistry (Paraffin) | |
LK2H10 + PHE5 | |
Unconjugated | |
Human pheochromocytoma | |
RUO | |
Primary | |
Concentrated |