missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Chromogranin A Ab-2, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Thermo Scientific™ Chromogranin A Ab-2, Mouse Monoclonal Antibody.

$247.18 - $778.98

Specifications

Antigen Chromogranin A Ab-2
Clone PHE5
Applications Immunofluorescence
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
MS381P Thermo Scientific™ Lab Vision™
MS381P
Purified Ab with BSA and Azide 1mL Each for $686.24
MS381P0 Thermo Scientific™ Lab Vision™
MS381P0
Purified Ab with BSA and Azide 100μL Each for $247.18
MS381P1 Thermo Scientific™ Lab Vision™
MS381P1
Purified Ab with BSA and Azide 500μL Each for $487.78
MS381P1ABX Thermo Scientific™ Lab Vision™
MS381P1ABX
Ab without BSA/Azide (Frozen) 100μL Each for $620.08
MS381PABX Thermo Scientific™ Lab Vision™
MS381PABX
Ab without BSA/Azide (Frozen) 200μL Each for $778.98
MS381R7 Thermo Scientific™ Lab Vision™
MS381R7
Ready-to-use for IHC (UVLP) 7mL Each for $359.02
Specifications

Description

Ensure accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Thermo Scientific™ Chromogranin A Ab-2, Mouse Monoclonal Antibody.

Chromogranin A (a protein of 439-amino acid which is encoded on chromosome 14) is present in neuroendocrine cells throughout the body, including the neuroendocrine cells of the large and small intestine, adrenal medulla and pancreatic islets. It is an excellent marker for carcinoid tumors, pheo-chromocytomas, paragangliomas, and other neuro-endocrine tumors. Coexpression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms.

Host Species: Mouse

Clone: PHE5

Isotype: IgG1

Species Reactivity: Human and Monkey. Does not react with rat. Others not known.

Immunogen: Human pheochromocytoma

Molecular Weight: 68-75 kDa

Positive Control: Pancreas

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Chromogranin A Ab-2
Immunofluorescence
Unlabeled
RUO
Primary
IgG1
PHE5
Monoclonal
Murine
Oncogenesis
Human pheochromocytoma