Please login to your online account to display your discounted pricing

Alfa Aesar™ Chromogranin A (324-337), Human

For cellular and molecular biology applications

Specifications

Product Type Chromogranin A (324-337), human
Cross Reactivity Human
Molecular Weight 1649.88
Storage Requirements -30° to -10°C
Sequence Trp-Ser-Lys-Met-Asp-Gln-Leu-Ala-Lys-Glu-Leu-Thr-Ala-Glu
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66711LB0

 
alfa aesar™
J66711LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66711MA

 
alfa aesar™
J66711MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Chromogranin A (324-337), human
Cross Reactivity Human
Molecular Weight 1649.88
Storage Requirements -30° to -10°C
Sequence Trp-Ser-Lys-Met-Asp-Gln-Leu-Ala-Lys-Glu-Leu-Thr-Ala-Glu
Format Solid
Common Name WE-14
Regulatory Status RUO
Purification Method HPLC