Please login to your online account to display your discounted pricing

Chromium(VI) Oxide, Reagent ACS +98%, ACROS Organics™


Quantity 1kg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC405230010

 View more versions of this product

Catalog No. AC405230010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Gently wash with plenty of soap and water. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Do not breathe dust/fume/gas/mist/vapors/spray. Avoid release to the environment.
Health Hazard 2 GHS H Statement
Fatal if inhaled. Causes damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause an allergic skin reaction. Suspected of damaging fertility. Very toxic to aquatic life with long lasting effects. May cause genetic defects. May cause cancer. May cause respiratory irritation. Fatal in contact with skin. May cause fire or explosion; strong oxidizer. Toxic if swallowed.
Purity Grade ACS Reagent Grade
Formula Weight 99.99
Physical Form Solid
Assay ≥98.0%
Quantity 1kg
Color Brown-red to purple
Merck Index 15, 2239
TSCA TSCA
Density 2.7000g/mL
RTECS Number GB6650000
EINECS Number 215-607-8
Fieser 01,144; 02,72; 03,54; 04,96; 05,140; 07,70; 09,115; 10,99; 13,338; 14,60
Assay Percent Range 98+%
Synonym Chromic anhydride
Sodium (Na) 0.2% max.
Chlorine (Cl) 50ppm max.
Sulfate 50ppm max.
CAS 1333-82-0
Linear Formula CrO3
Chemical Name or Material Chromium(VI) oxide
MDL Number MFCD00010952
Name Note 98+%
Molecular Formula CrO3
Packaging Glass bottle
Iron (Fe) 200ppm max.
Nitrate 500ppm max.
Aluminum (Al) 200ppm max.
Barium (Ba) 100ppm max.
Insoluble Matter 0.01% max.
Health Hazard 1 Danger

Specifications

Chemical Name or Material Chromium(VI) oxide
Name Note 98+%
CAS 1333-82-0
Purity Grade ACS Reagent Grade
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula CrO3
Linear Formula CrO3
Formula Weight 99.99
MDL Number MFCD00010952
Synonym Chromic anhydride
Accessories

Danger

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Gently wash with plenty of soap and water. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Do not breathe dust/fume/gas/mist/vapors/spray. Avoid release to the environment.

GHS H Statement
Fatal if inhaled. Causes damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause an allergic skin reaction. Suspected of damaging fertility. Very toxic to aquatic life with long lasting effects. May cause genetic defects. May cause cancer. May cause respiratory irritation. Fatal in contact with skin. May cause fire or explosion; strong oxidizer. Toxic if swallowed.

SDS & Safety Documents