missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chromium(VI) oxide, 98+%, ACS reagent, Acros Organics

$438.09 - $438.09

Chemical Identifiers

CAS 1333-82-0
Molecular Formula CrO3
Molecular Weight (g/mol) 99.99
MDL Number MFCD00010952
InChI Key WGLPBDUCMAPZCE-UHFFFAOYSA-N
Synonym anhydride chromique
PubChem CID 14915
ChEBI CHEBI:48240
IUPAC Name trioxochromium
SMILES O=[Cr](=O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC405235000 SDS View Documents Thermo Fisher Scientific
405235000
500g Glass bottle Each for $438.09
 
AC405230010 SDS View Documents Thermo Fisher Scientific
AC405230010
1kg Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Oxidation reagent
Specifications

Chemical Identifiers

1333-82-0
99.99
WGLPBDUCMAPZCE-UHFFFAOYSA-N
14915
trioxochromium
CrO3
MFCD00010952
anhydride chromique
CHEBI:48240
O=[Cr](=O)=O

Specifications

1333-82-0
98+%
CrO3
01,144; 02,72; 03,54; 04,96; 05,140; 07,70; 09,115; 10,99; 13,338; 14,60
anhydride chromique
WGLPBDUCMAPZCE-UHFFFAOYSA-N
trioxochromium
99.993
CHEBI:48240
Solid
ACS Reagent
2.7000g/mL
200ppm max.
Glass bottle
2.7
Brown-Red to Purple
500g
200ppm max.
CrO3
MFCD00010952
15, 2239
Solubility in water: 1660g/L (20°C). Other solubilities: soluble in sulfuric acid, soluble in alcohol and mineral acids
O=[Cr](=O)=O
100ppm max.
14915
99.99
≥98%
50ppm max.
0.01% max.
500ppm max.
0.2% max.
50ppm max.
196.0°C
Chromium(VI) oxide, 98+%
Safety and Handling

Safety and Handling

GHS H Statement
Fatal if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse

GHS Signal Word: Danger

EINECSNumber : 215-607-8

RTECSNumber : GB6650000

TSCA : TSCA