missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chromium(VI) Oxide, Reagent ACS +98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 405235000

 View more versions of this product

Catalog No. AC405235000


 

Specifications Description & Specifications

Specifications

Chromium(VI) oxide
1333-82-0
ACS Reagent Grade
≥98.0%
CrO3
99.99
01,144; 02,72; 03,54; 04,96; 05,140; 07,70; 09,115; 10,99; 13,338; 14,60
Chromic anhydride
50ppm max.
0.01% max.
500ppm max.
0.2% max.
Brown-red to purple
98+%
200ppm max.
Solid
98+%
CrO3
MFCD00010952
15, 2239
100ppm max.
2.7000g/mL
200ppm max.
Glass bottle
50ppm max.
500g

Chemical Identifiers

1333-82-0
CrO3
99.99
Chromic anhydride
≥98.0%
98+%
CrO3
MFCD00010952
Chromium(VI) oxide, 98+%
ACS Reagent Grade

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Fatal if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Suspected of damagin

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 215-607-8

RTECSNumber : GB6650000

TSCA : TSCA