missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chromium(VI) oxide, 99%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1252230

 View more versions of this product

Catalog No. AA1252230



Specifications

Specifications

Chromium(VI) oxide
99%
MFCD00010952
anhydride chromique
WGLPBDUCMAPZCE-UHFFFAOYSA-N
trioxochromium
14915
99.99
2.7 g/cm3
250°C (decomposition)
250g
1333-82-0
CrO3
Dark red crystals
Very soluble in water. Soluble in sulfuric,nitric acid,acetic acid,and acetone.
O=[Cr](=O)=O
99.993
CHEBI:48240
Crystalline
Hygroscopic
196°C

Chemical Identifiers

1333-82-0
99.993
WGLPBDUCMAPZCE-UHFFFAOYSA-N
14915
trioxochromium
CrO3
MFCD00010952
anhydride chromique
CHEBI:48240
O=[Cr](=O)=O
Safety and Handling

Safety and Handling

P201-P202-P210-P220-P221-P260-P262-P264b-P270-P271-P272-P280g-P281-P283-P284-P285-P302+P350-P304+P340-P305+P351+P338-P306+P360-P310-P330-P331-P333+P313-P353-P363-P370+P378q-P3

H271-H301-H310+H330-H314-H317-H334-H335-H340-H350-H361f-H372

DOTInformation : DOT Class: 5.1, Packaging Group: II

EINECSNumber : 215-607-8

RTECSNumber : GB6650000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents