missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Chromium(VI) oxide, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 1069136

 View more versions of this product

Catalog No. AA1069136



Specifications

Specifications

Chromium(VI) oxide
(metals basis)
MFCD00010952
14,2235
WGLPBDUCMAPZCE-UHFFFAOYSA-N
trioxochromium
14915
99.99
99.9%
Hygroscopic
Red
500g
1333-82-0
CrO3
UN1463
anhydride chromique
O=[Cr](=O)=O
99.993
CHEBI:48240
Crystalline
2.7g/mL
250°C (decomposition)
196°C

Chemical Identifiers

1333-82-0
99.993
WGLPBDUCMAPZCE-UHFFFAOYSA-N
14915
trioxochromium
CrO3
MFCD00010952
anhydride chromique
CHEBI:48240
O=[Cr](=O)=O
Safety and Handling

Safety and Handling

GHS H Statement
H271-H301-H310-H330-H334-H340-H350-H372-H361-H314-H318-H317
May cause fire or explosion
strong oxidizer.
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility or the unborn child.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.

GHS P Statement
P221-P283-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Take any precaution to avoid mixing with combustibles.
Wear fire/flame resistant/retardant clothing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see ... on this label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: CHROMIUM TRIOXIDE, ANHYDROUS

RTECSNumber : GB6650000

TSCA : TSCA

Documents