Please login to your online account to display your discounted pricing

Chromium(VI) oxide, 99.5%, ACROS Organics™


Quantity 2.5kg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC196610025

 View more versions of this product

Catalog No. AC196610025

  • / Each
 

Description & Specifications

Chemical Identifiers

Health Hazard 2 GHS H Statement
Fatal if inhaled. Causes damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause an allergic skin reaction. Suspected of damaging fertility. Very toxic to aquatic life with long lasting effects. May cause genetic defects. May cause cancer. May cause respiratory irritation. Fatal in contact with skin. May cause fire or explosion; strong oxidizer. Toxic if swallowed.
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Gently wash with plenty of soap and water. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Do not breathe dust/fume/gas/mist/vapors/spray. Avoid release to the environment.
Assay ≥99.4%
Physical Form Crystals or Granules
Formula Weight 99.99
Quantity 2.5kg
Color Dark red to purple
Density 2.7000g/mL
Merck Index 15, 2239
Fieser 01,144; 02,72; 03,54; 04,96; 05,140; 07,70; 09,115; 10,99; 13,338; 14,60
Synonym Chromic anhydride
CAS 1333-82-0
Assay Percent Range 99.5%
Health Hazard 1 Danger
Sulfate 500ppm max.
Linear Formula CrO3
Chemical Name or Material Chromium(VI) oxide
Molecular Formula CrO3
Packaging Glass bottle
MDL Number MFCD00010952

Specifications

Chemical Name or Material Chromium(VI) oxide
Assay ≥99.4%
Assay Percent Range 99.5%
Molecular Formula CrO3
Linear Formula CrO3
Formula Weight 99.99
MDL Number MFCD00010952
Synonym Chromic anhydride
Accessories

Danger

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Gently wash with plenty of soap and water. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Do not breathe dust/fume/gas/mist/vapors/spray. Avoid release to the environment.

GHS H Statement
Fatal if inhaled. Causes damage to organs through prolonged or repeated exposure. Causes severe skin burns and eye damage. May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause an allergic skin reaction. Suspected of damaging fertility. Very toxic to aquatic life with long lasting effects. May cause genetic defects. May cause cancer. May cause respiratory irritation. Fatal in contact with skin. May cause fire or explosion; strong oxidizer. Toxic if swallowed.

SDS & Safety Documents