Learn More
Chromium(VI) oxide, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 196610010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentSpecifications
Chromium(VI) oxide | |
1333-82-0 | |
Crystals or Granules | |
99.4% min. (Iodometry) | |
CrO3 | |
01,144; 02,72; 03,54; 04,96; 05,140; 07,70; 09,115; 10,99; 13,338; 14,60 | |
chromium trioxide, chromium vi oxide, monochromium trioxide, chromtrioxid, chromic trioxide, chromsaeureanhydrid, anhydride chromique, chromium 6+ oxide, chromium vi trioxide, chromium oxide | |
WGLPBDUCMAPZCE-UHFFFAOYSA-N | |
trioxochromium | |
14915 | |
99.99 | |
Glass bottle | |
2.7000g/mL |
196.0°C | |
Purple to Red | |
1 kg | |
CrO3 | |
MFCD00010952 | |
15, 2239 | |
Solubility in water: 1660g/L (20°C). Other solubilities: soluble in sulfuric acid, soluble in alcohol and mineral acids | |
O=[Cr](=O)=O | |
99.99 | |
CHEBI:48240 | |
99.5% | |
2.7 |
Chemical Identifiers
1333-82-0 | |
99.99 | |
WGLPBDUCMAPZCE-UHFFFAOYSA-N | |
14915 | |
trioxochromium |
CrO3 | |
MFCD00010952 | |
chromium trioxide, chromium vi oxide, monochromium trioxide, chromtrioxid, chromic trioxide, chromsaeureanhydrid, anhydride chromique, chromium 6+ oxide, chromium vi trioxide, chromium oxide | |
CHEBI:48240 | |
O=[Cr](=O)=O |
Safety and Handling
GHS H Statement
Fatal if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse
GHS Signal Word: Danger
EINECSNumber : 215-607-8
RUO – Research Use Only