missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hexavalent Chromium ICP Standard, 1000 ppm Cr6+ in 3% HNO3, Ricca Chemical
Supplier: Ricca Chemical Company PCR61KN100
Specifications
Chromium(VI) ICP Standard | |
Natural LDPE Bottle | |
94.0,4.22,0.23 | |
Yellow | |
Cr2H8N2O7 | |
[NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-] | |
252.06 | |
1 mL = 1 mg Cr |
7789-09-5,7697-37-2,7732-18-5 | |
100 mL | |
97.0,4.39,0.24 | |
1,000ppm Cr | |
JOSWYUNQBRPBDN-UHFFFAOYSA-P | |
diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium | |
24600 | |
Liquid |
Chemical Identifiers
7789-09-5 | |
252.06 | |
24600 | |
[NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-] |
Cr2H8N2O7 | |
JOSWYUNQBRPBDN-UHFFFAOYSA-P | |
diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium |