Please login to your online account to display your discounted pricing

Chromium Trioxide, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$183.54 - $601.19

Chemical Identifiers

Chemical Name or Material Chromium Trioxide
CAS 1333-82-0
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula CrO3
Formula Weight 99.99
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002603

 
avantor performance
163804
Wide Mouth Amber Glass Bottle 125g Each for $183.54

02002604

 
avantor performance
163801
Wide Mouth Amber Glass Bottle 500g Each for $601.19
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Physical Form Crystals
Formula Weight 99.99
CAS 1333-82-0
Chemical Name or Material Chromium Trioxide
Molecular Formula CrO3
Quantity 125g
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.70kg