missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chromium Trioxide ACS AR Crystal, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 1333-82-0
Physical Form Crystalline
Molecular Formula CrO3
Formula Weight 99.99
Chemical Name or Material Chromium Trioxide
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK2576A212 Avantor Performance
257605
Glass 2.5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

1333-82-0
CrO3
Chromium Trioxide
Crystalline
99.99
ACS Grade

Specifications

99.99
Crystalline
Glass
Chromium Trioxide
2.5kg
CrO3
1333-82-0
ACS Grade