Please login to your online account to display your discounted pricing

Alfa Aesar™ Chromium sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.95% (metals basis)

$775.99 - $775.99

Chemical Identifiers

Chemical Name or Material Chromium sputtering target
CAS 7440-47-3
Name Note 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
MDL Number MFCD00010944
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40845KS

  SDS
alfa aesar™
40845KS
1 each Each for $775.99
Description & Specifications

Specifications

CAS 7440-47-3
Assay 99.95%
Chemical Name or Material Chromium sputtering target
TSCA TSCA
Assay Percent Notes (metals basis)
Quantity 1 each
Name Note 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
MDL Number MFCD00010944
Odor Odorless