missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Chromium sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.95% (metals basis), Thermo Scientific™
Supplier: Thermo Scientific Chemicals 40843KS
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Chromium sputtering target | |
7440-47-3 | |
Cr | |
1 Ea. | |
Insoluble in water. | |
[Cr] | |
52.00 | |
CHEBI:28073 | |
Odorless |
50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick | |
(metals basis) | |
MFCD00010944 | |
chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159 | |
VYZAMTAEIAYCRO-UHFFFAOYSA-N | |
chromium | |
23976 | |
99.95% |
Chemical Identifiers
7440-47-3 | |
52.00 | |
VYZAMTAEIAYCRO-UHFFFAOYSA-N | |
23976 | |
chromium |
Cr | |
MFCD00010944 | |
chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159 | |
CHEBI:28073 | |
[Cr] |
Safety and Handling
EINECSNumber : 231-157-5
TSCA : Yes
Recommended Storage : Ambient temperatures