Please login to your online account to display your discounted pricing

Alfa Aesar™ Chromium sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.95% (metals basis)

Magnetron sputtering source

$577.23 - $577.23

Chemical Identifiers

Chemical Name or Material Chromium sputtering target
CAS 7440-47-3
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
MDL Number MFCD00010944
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40842KS

  SDS
alfa aesar™
40842KS
1 each Each for $577.23
Description & Specifications

Specifications

Chemical Name or Material Chromium sputtering target
TSCA TSCA
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Assay 99.95%
Assay Percent Notes (metals basis)
MDL Number MFCD00010944
CAS 7440-47-3
Odor Odorless
Quantity 1 each