Please login to your online account to display your discounted pricing

Alfa Aesar™ Chromium silicon monoxide, 99.9% (metals basis)

Chemical Identifiers

Chemical Name or Material Chromium silicon monoxide
CAS 37264-20-3
MDL Number MFCD02091702
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3566709

  SDS
alfa aesar™
3566709
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3566718

  SDS
alfa aesar™
3566718
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Health Hazard 3 GHS P Statement
P284a-P261-P280-P342+P311-P363-P501a
Wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Chromium silicon monoxide
TSCA TSCA
Mesh Size −8+20 Mesh
Assay 99.9%
Assay Percent Notes (metals basis)
MDL Number MFCD02091702
CAS 37264-20-3
Quantity 10g
Avg. Mol. Wt. or Mol. Wt. Range Cr:SiO; 50:50 wt%
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P342+P311-P363-P501a
Wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.