missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chromium silicon monoxide, 99.8+% (metals basis), Thermo Scientific™

For vacuum deposition

$281.23 - $939.05

Chemical Identifiers

CAS 56488-64-3
MDL Number MFCD02091702
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3637814
View Documents Promotion Details
Thermo Scientific Chemicals
3637814
25 g
Each for $281.23
 
AA3637822
View Documents Promotion Details
Thermo Scientific Chemicals
3637822
100 g
Each for $939.05
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

56488-64-3
MFCD02091702

Specifications

56488-64-3
25 g
Insoluble in water.
Chromium silicon monoxide
Sintered Pieces
MFCD02091702
≥99.8%
(metals basis)
Safety and Handling

Safety and Handling

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c

H317-H334

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'