Please login to your online account to display your discounted pricing

Alfa Aesar™ Chromium silicide sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.5% (metals basis)

Chemical Identifiers

Chemical Name or Material Chromium silicide sputtering target
Name Note 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
Formula Weight 108.17
MDL Number MFCD00168070
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41063KS

 
alfa aesar™
41063KS
1 each CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 108.17
Health Hazard 3 GHS P Statement
P284a-P261-P280-P342+P311-P363-P501a
Wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Chemical Name or Material Chromium silicide sputtering target
TSCA TSCA
Name Note 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
Assay 99.5%
Assay Percent Notes (metals basis)
MDL Number MFCD00168070
Quantity 1 each
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P342+P311-P363-P501a
Wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.