Please login to your online account to display your discounted pricing

Alfa Aesar™ Chromium silicide sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.5% (metals basis)

Magnetron sputtering source

Chemical Identifiers

Chemical Name or Material Chromium silicide sputtering target
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Formula Weight 108.17
MDL Number MFCD00168070
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41060KS

  SDS
alfa aesar™
41060KS
1 each CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 108.17
Health Hazard 3 GHS P Statement
P284a-P261-P280-P342+P311-P363-P501a
Wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1 each
Chemical Name or Material Chromium silicide sputtering target
Health Hazard 1 Danger
TSCA TSCA
Assay 99.5%
Assay Percent Notes (metals basis)
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Health Hazard 2 GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
MDL Number MFCD00168070
Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P342+P311-P363-P501a
Wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.