missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chromium silicide, >99% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1215118*

 View more versions of this product

Catalog No. AA1215118



Specifications

Specifications

Chromium silicide
>95%
MFCD00168070
chromsilicid, chromium silicide, bis $l^ 3
[Si]#[Cr]#[Si]
108.166
108.17
Odorless
50g
12018-09-6
CrSi2
Gray powder
CHXGWONBPAADHP-UHFFFAOYSA-N
bis($l^{3}-silanylidyne)chromium
45051936
Powder
Gray

Chemical Identifiers

12018-09-6
108.166
CHXGWONBPAADHP-UHFFFAOYSA-N
45051936
[Si]#[Cr]#[Si]
CrSi2
MFCD00168070
chromsilicid, chromium silicide, bis $l^ 3
bis($l^{3}-silanylidyne)chromium
Safety and Handling

Safety and Handling

H334-H315-H319-H317-H335 : May cause allergy or asthma symptoms or breathing difficulties if inhaled. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. May cause respiratory irritation.

EINECSNumber : 234-633-0

Documents