Learn More
Chromium silicide, 99+% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01215130
Description
Chromium silicide is applied in production of ceramic materials. It can also produce an oxidation-resistant surface in a low pressure plasma spraying.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Chromium silicide | |
12018-09-6 | |
Powder | |
>95% | |
MFCD00168070 | |
Insoluble in water. | |
[Si]#[Cr]#[Si] | |
108.166 | |
108.17 | |
Moisture sensitive |
1490°C | |
Gray | |
250 g | |
CrSi2 | |
chromsilicid, chromium silicide, bis $l^ 3 | |
CHXGWONBPAADHP-UHFFFAOYSA-N | |
bis($l^{3}-silanylidyne)chromium | |
45051936 | |
Odorless | |
Gray powder |
Chemical Identifiers
12018-09-6 | |
108.166 | |
CHXGWONBPAADHP-UHFFFAOYSA-N | |
45051936 | |
[Si]#[Cr]#[Si] |
CrSi2 | |
MFCD00168070 | |
chromsilicid, chromium silicide, bis $l^ 3 | |
bis($l^{3}-silanylidyne)chromium |
Safety and Handling
P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P333+P313-P342+P311-P362-P501c
H315-H317-H319-H334-H335
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 234-633-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only