Please login to your online account to display your discounted pricing

Chromium, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical

For atomic absorption. (1mL = 1mg Cr) Solute: Potassium Dichromate Solvent: Distilled H2O

$86.41 - $119.65

Chemical Identifiers

Chemical Name or Material Chromium, Reference Standard Solution
Name Note 1000ppm ±1%
CAS 7778-50-9
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00010944
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SC192-100

  SDS
fisher scientific company
SC192100
Poly Bottle 100mL Each for $86.41
Case of 6 Each for $518.46

SC192-500

  SDS
fisher scientific company
SC192500
Poly Bottle 500mL Each for $119.65
Case of 6 Each for $717.90
Description & Specifications

Specifications

Purity Grade Certified Grade
Melting Point 0°C
Boiling Point 100°C
Quantity 100mL
Color Orange
Physical Form Liquid
Vapor Pressure 14mmHg at 20°C
Viscosity [NULL]
Density 1.0g/cm³
pH 7
Chemical Name or Material Chromium, Reference Standard Solution
Concentration or Composition (by Analyte or Components) 1000ppm ±1%
CAS 7732-18-5
Packaging Poly Bottle
MDL Number MFCD00010944
CAS 7778-50-9
ChemAlert Storage Symbol Gray
Traceability to NIST 136f POTASSIUM DICHROMATE
Concentration 990 to 1010ppm
Name Note 1000ppm ±1%
Safety and Handling