missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chromium powder, 99.99% (metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1045222

 View more versions of this product

Catalog No. AA1045222



Specifications

Specifications

Chromium
7440-47-3
99.996% (Metals basis)
MFCD00010944
VYZAMTAEIAYCRO-UHFFFAOYSA-N
chromium
51.996
CHEBI:28073
0 hPa
100g
Gray powder
Cr
ccris 159
[Cr]
Odorless
23976
Powder
Gray

Chemical Identifiers

7440-47-3
51.996
VYZAMTAEIAYCRO-UHFFFAOYSA-N
23976
chromium
Cr
MFCD00010944
ccris 159
CHEBI:28073
[Cr]
Safety and Handling

Safety and Handling

H351: Suspected of causing cancer.

EINECSNumber : 231-157-5

Documents