missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chromium Powder, -60 mesh, 99.99% (Metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 4455636

 View more versions of this product

Catalog No. AA4455636



Specifications

Specifications

Chromium
7440-47-3
99.99% (Metals basis)
MFCD00010944
ccris 159
[Cr]
Odorless
23976
Powder
500g
Gray powder
Cr
-60 Mesh
VYZAMTAEIAYCRO-UHFFFAOYSA-N
chromium
51.996
CHEBI:28073
Gray

Chemical Identifiers

7440-47-3
51.996
VYZAMTAEIAYCRO-UHFFFAOYSA-N
23976
chromium
Cr
MFCD00010944
ccris 159
CHEBI:28073
[Cr]
Safety and Handling

Safety and Handling

P210-P240-P241-P280-P370+P378q

H228

DOTInformation : Hazard Class: 4.1; Packaging Group: III

EINECSNumber : 231-157-5

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents